Merck
  • Zinc oxide nanoparticles induced oxidative stress in mouse bone marrow mesenchymal stem cells.
Zinc oxide nanoparticles induced oxidative stress in mouse bone marrow mesenchymal stem cells.
Toxicology mechanisms and methods (2014-08-21)
S Syama, P J Sreekanth, H K Varma, P V Mohanan
ABSTRACT
Engineered nanoparticles are developed for various applications in industrial, electrical, agricultural, pharmaceutical and medical fields due to their unique properties. Nanoparticles such as TiO(2) and ZnO are widely used in cosmetics for UV protection. The toxicological investigations of ZnO NPs are highly recommended because of the increasing use in various industrial and consumer products. The toxic potential of ZnO NPs was assumed to be caused by the release of free Zn+ ions in the medium. Many of the in vivo studies suggest the toxic nature of ZnO NPs, the in vitro studies are certainly important to elucidate the mechanism of toxicity. This study examined the toxicity of ZnO NPs with the average size of 6-8 nm on the isolated mice bone marrow mesenchymal stem cells. The study focuses on the cytotoxicity and oxidative stress-mediated cellular responses upon exposure to ZnO NPs. The results indicated that the exposure to ZnO NPs significantly affects cellular viability in a dose-dependent manner. Formation of reactive oxygen species (ROS) was found to be the mechanism of cellular toxicity. The release of Zn(+) ions from the nanoparticles, due to the instability of ZnO NPs in the acidic compartment of lysosomes, also increases the ROS generation. In addition to increased ROS production, damage of lysosomal membrane and the activation of executioner caspase-3 and caspase-7 were observed, which eventually ends in apoptosis.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%