Merck
  • SrO- and MgO-doped microwave sintered 3D printed tricalcium phosphate scaffolds: mechanical properties and in vivo osteogenesis in a rabbit model.
SrO- and MgO-doped microwave sintered 3D printed tricalcium phosphate scaffolds: mechanical properties and in vivo osteogenesis in a rabbit model.
Journal of biomedical materials research. Part B, Applied biomaterials (2014-07-22)
Solaiman Tarafder, William S Dernell, Amit Bandyopadhyay, Susmita Bose
ABSTRACT
The presence of interconnected macro pores allows guided tissue regeneration in tissue engineering scaffolds. However, highly porous scaffolds suffer from having poor mechanical strength. Previously, we showed that microwave sintering could successfully be used to improve mechanical strength of macro porous tricalcium phosphate (TCP) scaffolds. This study reports the presence of SrO and MgO as dopants in TCP scaffolds improves mechanical and in vivo biological performance. We have used direct three dimensional printing (3DP) technology for scaffold fabrication. These 3DP scaffolds possessed multiscale porosity, that is, 3D interconnected designed macro pores along with intrinsic micro pores. A significant increase in mechanical strength, between 37 and 41%, was achieved due to SrO and MgO doping in TCP as compared with pure TCP. Maximum compressive strengths of 9.38 ± 1.86 MPa and 12.01 ± 1.56 MPa were achieved by conventional and microwave sintering, respectively, for SrO-MgO-doped 3DP scaffolds with 500 μm designed pores. Histomorphological and histomorphometric analysis revealed a significantly higher osteoid, bone and haversian canal formation induced by the presence of SrO and MgO dopants in 3DP TCP as compared with pure TCP scaffolds when tested in rabbit femoral condyle defect model. Increased osteon and thus enhanced network of blood vessel formation, and osteocalcin expression were observed in the doped TCP scaffolds. Our results show that these 3DP SrO-MgO-doped TCP scaffolds have the potential for early wound healing through accelerated osteogenesis and vasculogenesis.
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Pricing and availability is not currently available.
Sigma-Aldrich
Strontium oxide, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Pricing and availability is not currently available.
Supelco
3,5,6-Trichloro-2-pyridinol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, light, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Pricing and availability is not currently available.