Merck
  • Procedure used for denuding pig oocytes influences oocyte damage, and development of in vitro and nuclear transfer embryos.
Procedure used for denuding pig oocytes influences oocyte damage, and development of in vitro and nuclear transfer embryos.
Animal reproduction science (2014-12-10)
Tao Lin, Yun Fei Diao, Hwa Sik Choi, Reza K Oqani, Jung Won Kang, Jae Eun Lee, Dong Il Jin
ABSTRACT
The effects of different denuding procedures used during the in vitro culture of porcine embryos on oocyte damage and aspects of porcine embryo development were investigated in a series of studies. Oocytes were denuded by vortexing or pipetting after 44h in vitro maturation (IVM) or pre-denuded after 22h IVM. The total oocyte death rate was significantly (P<0.05) higher for pre-denuded (27.3±1.4%) than for vortexed (20.3±1.2%) or pipetted (16.2±2.2%) oocytes. There was no significant difference between the treatments in the percentage of oocytes that extruded the first polar body. The type I cortical granule distribution (reflecting complete maturity) and normal spindle formation rates were significantly lower in the pre-denuding than in the vortexing and pipetting treatments. Blastocyst formation rates were significantly lower for the pre-denuding treatment in PA (25.7±4.5%) and IVF (6.1±1.5%) culture than in the vortexing (PA 42.0±4.5%; IVF 11.2±0.5%) and pipetting (PA 43.4±3.1%; IVF 9.4±1.6%) treatments. The proportion of oocytes developing to blastocysts in SCNT culture was not significantly different between treatments ranging from 9.9±1.8% for pre-denuding to 12.3±2.7% for vortexing. No significant differences in apoptosis or embryonic fragmentation were observed. This study shows that the denuding procedure used for porcine oocytes during the in vitro production of embryos can significantly affect oocyte damage, spindle patterns, oocyte maturation, embryo development but not embryonic apoptosis or the frequency of fragmentation.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture