Merck
  • Sex-dependent decrease of sphingomyelinase activity during alcohol withdrawal treatment.
Sex-dependent decrease of sphingomyelinase activity during alcohol withdrawal treatment.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2014-07-01)
Christiane Mühle, Violina Amova, Teresa Biermann, Kristina Bayerlein, Tanja Richter-Schmidinger, Thomas Kraus, Martin Reichel, Erich Gulbins, Johannes Kornhuber
ABSTRACT
In vitro and in vivo studies have demonstrated the role of the acid sphingomyelinase (ASM) in pathophysiological processes and alterations in response to ethanol exposure. Cellular and plasmatic ASM activities are increased in male alcohol dependent patients and decrease during physical withdrawal. Here, we analyzed the time course of ASM in male and also female acutely intoxicated patients during alcohol withdrawal and compared the activity levels to those under long-term maintenance treatment. Craving and further psychometric parameters were assessed by questionnaires. The gradual decrease of serum ASM was confirmed in males (p<0.001) and continued to lower activities in long-term patients (p=0.001). The trend was similar in females (p=0.178), although the initial enzyme activities were significantly lower (p=0.035). ASM activity strongly correlated with the body mass index in males. The initial ASM activity and its decline during the first two days were associated with the improvement in scores for the Beck depression inventory, the obsessive compulsive drinking and the withdrawal syndrome scales. These data support the potential of ASM as a biomarker for the course of withdrawal therapy in males and provide the first associations of this enzyme with psychological variables such as craving and depression.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%