Merck
  • Growth suppression and cell death by pyridoxal is dependent on p53 in the human breast cancer cell line MCF-7.
Growth suppression and cell death by pyridoxal is dependent on p53 in the human breast cancer cell line MCF-7.
Bioscience, biotechnology, and biochemistry (2015-01-22)
Masaki Minamino, Tatsuzo Oka, Hiroaki Kanouchi
ABSTRACT
Vitamin B6 compound, pyridoxine (PN), has shown antitumor action. Our previous experiments showed that PN induces expression of insulin-like growth factor binding protein-3 to arrest proliferation and induce cell death. This induction is inhibited by the p53-specific inhibitor pifithrin-α. Here, we report that another B6 compound, pyridoxal (PL), strongly inhibited MCF-7 cell growth compared to PN. PL induced the G0/G1 arrest and the accumulation of subG1 population. Although p53 mRNA was not changed by PL, 0.5 mM PL increased the protein level in MCF-7 cells. The cell growth suppression by 0.5 mM PL did not occur when p53 expression was knocked down using siRNA. Together, these data suggest that PL accumulate p53 and PL-induced cell growth suppression is dependent on p53 in MCF-7 breast cancer cells.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-1, purified from hybridoma cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%