Merck
  • Dynamics of the two heterochromatin types during imprinted X chromosome inactivation in vole Microtus levis.
Dynamics of the two heterochromatin types during imprinted X chromosome inactivation in vole Microtus levis.
PloS one (2014-02-08)
Evgeniya A Vaskova, Elena V Dementyeva, Alexander I Shevchenko, Sophia V Pavlova, Elena V Grigor'eva, Antonina I Zhelezova, John L Vandeberg, Suren M Zakian
ABSTRACT
In rodent female mammals, there are two forms of X-inactivation - imprinted and random which take place in extraembryonic and embryonic tissues, respectively. The inactive X-chromosome during random X-inactivation was shown to contain two types of facultative heterochromatin that alternate and do not overlap. However, chromatin structure of the inactive X-chromosome during imprinted X-inactivation, especially at early stages, is still not well understood. In this work, we studied chromatin modifications associated with the inactive X-chromosome at different stages of imprinted X-inactivation in a rodent, Microtus levis. It has been found that imprinted X-inactivation in vole occurs in a species-specific manner in two steps. The inactive X-chromosome at early stages of imprinted X-inactivation is characterized by accumulation of H3K9me3, HP1, H4K20me3, and uH2A, resembling to some extent the pattern of repressive chromatin modifications of meiotic sex chromatin. Later, the inactive X-chromosome recruits trimethylated H3K27 and acquires the two types of heterochromatin associated with random X-inactivation.
Sigma-Aldrich
Anti-HP1α Antibody, clone15.19s2, clone 15.19s2, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
FGF-4 human, Animal-component free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Pricing and availability is not currently available.