Merck
  • Comparative transcriptome analyses between a spontaneous late-ripening sweet orange mutant and its wild type suggest the functions of ABA, sucrose and JA during citrus fruit ripening.
Comparative transcriptome analyses between a spontaneous late-ripening sweet orange mutant and its wild type suggest the functions of ABA, sucrose and JA during citrus fruit ripening.
PloS one (2015-01-01)
Ya-Jian Zhang, Xing-Jian Wang, Ju-Xun Wu, Shan-Yan Chen, Hong Chen, Li-Jun Chai, Hua-Lin Yi
ABSTRACT
A spontaneous late-ripening mutant of 'Jincheng' (C. sinensis L. Osbeck) sweet orange exhibited a delay of fruit pigmentation and harvesting. In this work, we studied the processes of orange fruit ripening through the comparative analysis between the Jincheng mutant and its wild type. This study revealed that the fruit quality began to differ on 166th days after anthesis. At this stage, fruits were subjected to transcriptome analysis by RNA sequencing. 13,412 differentially expressed unigenes (DEGs) were found. Of these unigenes, 75.8% were down-regulated in the wild type, suggesting that the transcription level of wild type was lower than that of the mutant during this stage. These DEGs were mainly clustered into five pathways: metabolic pathways, plant-pathogen interaction, spliceosome, biosynthesis of plant hormones and biosynthesis of phenylpropanoids. Therefore, the expression profiles of the genes that are involved in abscisic acid, sucrose, and jasmonic acid metabolism and signal transduction pathways were analyzed during the six fruit ripening stages. The results revealed the regulation mechanism of sweet orange fruit ripening metabolism in the following four aspects: First, the more mature orange fruits were, the lower the transcription levels were. Second, the expression level of PME boosted with the maturity of the citrus fruit. Therefore, the expression level of PME might represent the degree of the orange fruit ripeness. Third, the interaction of PP2C, PYR/PYL, and SnRK2 was peculiar to the orange fruit ripening process. Fourth, abscisic acid, sucrose, and jasmonic acid all took part in orange fruit ripening process and might interact with each other. These findings provide an insight into the intricate process of sweet orange fruit ripening.
Sigma-Aldrich
Sucrose, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on 23 March 2025
MXP 3,757.00