Merck
  • Yttrium oxide/gadolinium oxide-modified platinum nanoparticles as cathodes for the oxygen reduction reaction.
Yttrium oxide/gadolinium oxide-modified platinum nanoparticles as cathodes for the oxygen reduction reaction.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-05-14)
Yun Luo, Aurélien Habrioux, Laura Calvillo, Gaetano Granozzi, Nicolas Alonso-Vante
ABSTRACT
Rare-earth-element (Y, Gd) modified Pt nanoparticles (NPs) supported on a carbon substrate (Vulcan XC-72) are synthesized via a water-in-oil chemical route. In both cases, X-ray diffraction (XRD) measurements show the non-formation of an alloyed material. Photoemission spectroscopy (XPS) results reveal that Y and Gd are oxidized. Additionally, no evidence of an electronic modification of Pt can be brought to light. Transmission electron microscopy (TEM) studies indicate that Pt-Y(2)O(3) and Pt-Gd(2)O(3) particles are well dispersed on the substrate-and that their average particle sizes are smaller than the Pt-NP sizes. The catalytic activity of the Pt-Y(2)O(3)/C and Pt-Gd(2)O(3)/C catalysts towards the oxygen reduction reaction (ORR) is studied in a 0.5 M H(2)SO(4) electrolyte. The surface and mass specific activities of the Pt-Y(2)O(3)/C catalyst towards the ORR at 0.9 V (vs. the reversible hydrogen electrode, RHE) are (54.3±1.2) μA cm(-2)(Pt) and MA=(23.1±0.5) mA mg(-1)(Pt), respectively. These values are 1.3-, and 1.6-fold higher than the values obtained with a Pt/C catalyst. Although the as-prepared Pt-Gd(2)O(3)/C catalyst has a lower catalytic activity for the ORR compared to Pt/C, the heat-treated sample shows a surface specific activity of about (53.0±0.7) μA cm(-2) Pt , and a mass specific activity (MA) of about (18.2±0.5) mA mg(-1) Pt at 0.9 V (vs. RHE). The enhancement of the ORR kinetics on the Pt-Y(2)O(3)/C and heat-treated Pt-Gd(2)O(3)/C catalysts could be associated with the formation of platinum NPs presenting modified surface properties.
Product Description
Supelco
Heptane, analytical standard
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
VenPure® SF, powder
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Supelco
Gadolinium(III) chloride solution, NMR reference standard, water 1 %, DSS 0.1 %, 0.1 mg/mL in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Yttrium(III) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Yttrium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis