Merck
  • Preparation of chiral ligands connected with quaternary ammonium group for recyclable catalytic asymmetric transfer hydrogenation in ionic liquid.
Preparation of chiral ligands connected with quaternary ammonium group for recyclable catalytic asymmetric transfer hydrogenation in ionic liquid.
Chemical & pharmaceutical bulletin (2015-03-12)
Hitomi Uchimoto, Tomoko Tsuji, Ikuo Kawasaki, Kenji Arimitsu, Hiroyuki Yasui, Masayuki Yamashita, Shunsaku Ohta, Kiyoharu Nishide
ABSTRACT
Reuse of chiral ruthenium catalyst in catalytic asymmetric transfer hydrogenation (CATH) has attracted attention from economic and environmental viewpoints, and reactions using ionic liquids (ILs) as solvent are recognized as one of the most useful methods for reuse of the catalyst. We synthesized (1S,2S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine (TsDPEN) derivatives with various ionic moieties, and investigated the effect of their structure with respect to catalytic ability and recyclability in CATH with ILs. Ligand 3a having an imidazolium group showed the best results, and significant differences were observed depending on the structure of the ionic moiety or the length of the alkyl chain connecting the ligand site and the ionic moiety. Among various prochiral ketones used as substrates at various cycles, 3a showed a relatively good result.
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis