Merck
  • Structure-function relationship of thiazolide-induced apoptosis in colorectal tumor cells.
Structure-function relationship of thiazolide-induced apoptosis in colorectal tumor cells.
ACS chemical biology (2014-05-09)
Anette Brockmann, Tobias Strittmatter, Sarah May, Kerstin Stemmer, Andreas Marx, Thomas Brunner
ABSTRACT
Thiazolides are a novel class of anti-infectious agents against intestinal intracellular and extracellular protozoan parasites, bacteria, and viruses. While the parent compound nitazoxanide (NTZ; 2-(acetolyloxy)-N-(5-nitro-2-thiazolyl)benzamide) has potent antimicrobial activity, the bromo-thiazolide RM4819 (N-(5-bromothiazol-2-yl)-2-hydroxy-3-methylbenzamide) shows only reduced activity. Interestingly, both molecules are able to induce cell death in colon carcinoma cell lines, indicating that the molecular target in intestinal pathogens and in colon cancer cells is different. The detoxification enzyme glutathione S-transferase of class Pi 1 (GSTP1) is frequently overexpressed in various tumors, including colon carcinomas, and limits the efficacy of antitumor chemotherapeutic drugs due to its detoxifying activities. In colorectal tumor cells RM4819 has been shown to interact with GSTP1, and GSTP1 enzymatic activity is required for thiazolide-induced apoptosis. At present it is unclear which molecular structures of RM4819 are required to interact with GSTP1 and to induce cell death in colon carcinoma cell lines. Here, we demonstrate that novel thiazolide derivatives with variation in their substituents of the benzene ring do not significantly affect apoptosis induction in Caco-2 cells, whereas removal of the bromide atom on the thiazole ring leads to a strong reduction of cell death induction in colon cancer cells. We further show that active thiazolides require caspase activation and GSTP1 expression in order to induce apoptosis. We demonstrate that increased glutathione (GSH) levels sensitize colon cancer cells to thiazolides, indicating that both GSTP1 enzymatic activity as well as GSH levels are critical factors in thiazolide-induced cell death.
Sigma-Aldrich
MISSION® esiRNA, targeting human GSTP1
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)