Merck
  • A folate-integrated magnetic polymer micelle for MRI and dual targeted drug delivery.
A folate-integrated magnetic polymer micelle for MRI and dual targeted drug delivery.
Nanoscale (2014-08-07)
Lijiao Ao, Bi Wang, Peng Liu, Liang Huang, Caixia Yue, Duyang Gao, Chunlei Wu, Wu Su
ABSTRACT
This paper devotes a novel micellar structure for cancer theranostics by incorporating magnetic and therapeutic functionalities into a natural sourced targeting polymer vehicle. Heparin-folic acid micelles taking advantage of both excellent loading capability and cancer targeting ability have been employed to simultaneously incorporate superparamagnetic iron oxide nanoparticles (SPIONs) and doxorubicin through an ultrasonication-assisted microemulsion method. In this system, folic acids not only take the responsibility of micelle construction, but also facilitate cellular uptake due to their specific reorganization by MCF-7 cells over-expressing folate receptors. The obtained micelles exhibit good colloidal stability, a high magnetic content, considerable drug loading and sustained in vitro drug release. These clustered SPIONs exhibited high r2 relaxivity (243.65 mM(-1) s(-1)) and further served as efficient probes for MR imaging. Notably, the transport efficiency of these micelles could be significantly improved under an external magnetic field, owing to their quick magnetic response. As a result, the as-proposed micelle shows great potential in multimodal theranostics, including active targeting, MRI diagnosis and drug delivery.
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Dibenzyl ether, purum, ≥98.0% (GC)
Sigma-Aldrich
Benzyl ether, ≥98%, FCC, FG
Sigma-Aldrich
1,2-Dodecanediol, 90%
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Tributylamine, puriss. plus, ≥99.5% (GC)
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Oleylamine, technical grade, 70%
Supelco
Oleic acid, Selectophore, for ion-selective electrodes, ≥99%
Sigma-Aldrich
Tributylamine, puriss. p.a., ≥99.0% (GC)
Oleic acid, European Pharmacopoeia (EP) Reference Standard