Merck
Biodegradation of methyl tert-butyl ether by a bacterial pure culture.
Applied and environmental microbiology (1999-11-05)
J R Hanson, C E Ackerman, K M Scow
ABSTRACT
A bacterial strain, PM1, which is able to utilize methyl tert-butyl ether (MTBE) as its sole carbon and energy source, was isolated from a mixed microbial consortium in a compost biofilter capable of degrading MTBE. Initial linear rates of MTBE degradation by 2 x 10(6) cells ml(-1) were 0.07, 1.17, and 3.56 microg ml(-1) h(-1) for initial concentrations of 5, 50, and 500 microg MTBE ml(-1), respectively. When incubated with 20 microg of uniformly labeled [(14)C]MTBE ml(-1), strain PM1 converted 46% to (14)CO(2) and 19% to (14)C-labeled cells within 120 h. This yield is consistent with the measurement of protein accumulation at different MTBE concentrations from which was estimated a biomass yield of 0.18 mg of cells mg MTBE(-1). Strain PM1 was inoculated into sediment core material collected from a contaminated groundwater plume at Port Hueneme, California, in which there was no evidence of MTBE degradation. Strain PM1 readily degraded 20 microg of MTBE ml(-1) added to the core material. The rate of MTBE removal increased with additional inputs of 20 microg of MTBE ml(-1). These results suggest that PM1 has potential for use in the remediation of MTBE-contaminated environments.
Product Description
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)