Merck
  • Assessment of morphological changes of Clostridium acetobutylicum by flow cytometry during acetone/butanol/ethanol extractive fermentation.
Assessment of morphological changes of Clostridium acetobutylicum by flow cytometry during acetone/butanol/ethanol extractive fermentation.
Biotechnology letters (2014-10-30)
Helena González-Peñas, Thelmo Alejandro Lu-Chau, Maria Teresa Moreira, Juan Manuel Lema
ABSTRACT
Acetone/butanol/ethanol (ABE) fermentation by Clostridium acetobutylicum was investigated in extractive fed-batch experiments. In conventional fermentations, metabolic activity ceases when a critical threshold products concentration is reached (~21.6 g solvents l(-1)). Solvents production was increased up to 36.6 and 37.2 g l(-1), respectively, using 2-butyl-1-octanol (aqueous to organic ratio: 1:0.25 v/v) and pomace olive oil (1:1 v/v) as extraction solvents. The morphological changes of different cell types were monitored and quantified using flow cytometry. Butanol production in extractive fermentations with pomace olive oil was achieved mainly by vegetative cells, whereas the percentage of sporulating cells was lower than 10%.
Sigma-Aldrich
2-Butyl-1-octanol, 95%
Pricing and availability is not currently available.