Merck
  • Molecularly imprinted polymers with synthetic dummy template for simultaneously selective removal and enrichment of ginkgolic acids from Ginkgo biloba L. leaves extracts.
Molecularly imprinted polymers with synthetic dummy template for simultaneously selective removal and enrichment of ginkgolic acids from Ginkgo biloba L. leaves extracts.
Journal of chromatography. A (2014-12-03)
Wenhua Ji, Xiuli Ma, Hongkai Xie, Lingxiao Chen, Xiao Wang, Hengqiang Zhao, Luqi Huang
ABSTRACT
Dummy molecularly imprinted polymers (DMIPs) for simultaneously selective removal and enrichment of ginkgolic acids (GAs) during the processing of Ginkgo biloba leaves have been prepared. Two dummy template molecule with similar structural skeleton to GAs, 6-methoxysalicylic acid (MOSA, DT-1) and 6-hexadecyloxysalicylic acid (HOSA, DT-2), have been designed and synthesized. The performance of the DMIPs and NIPs were evaluated including selective recognition capacity, adsorption isotherm, and adsorption kinetics. The selective recognition capacity of the three GAs with four analogues on the sorbents illustrated that the DMIPs sorbents have high specificity for GAs. An efficient method based on DMIP-HOSA coupled with solid-phase extraction (SPE) was developed for simultaneously selective removal and enrichment of ginkgolic acids (GAs) during the processing of Ginkgo biloba leaves. The method showed excellent recoveries (82.5-88.7%) and precision (RSD 0.5-2.6%, n=5) for licorice extracts, Gastrodia elata extracts and pepper extracts spiked at three concentration levels each (50, 100, 200 μg mL(-1)). The results indicated that GAs and standardized Ginkgo biloba leaves extracts could be obtained simultaneously through the DMIP-SPE.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thionyl chloride, reagent grade, 97%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Dimethyl sulfate, ≥99.5%
Sigma-Aldrich
Diisopropyl azodicarboxylate, 98%
Sigma-Aldrich
Dimethyl sulfate, ≥99%
Supelco
Acetic acid, analytical standard
Supelco
1,2-Dimethoxyethane, analytical standard
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free
Sigma-Aldrich
Dimethyl sulfate, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
1,2-Dimethoxyethane, suitable for HPLC, 99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
1,2-Dimethoxyethane, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%