Merck
  • PICK1 mediates synaptic recruitment of AMPA receptors at neurexin-induced postsynaptic sites.
PICK1 mediates synaptic recruitment of AMPA receptors at neurexin-induced postsynaptic sites.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-11-14)
Junyu Xu, Chuen Kam, Jian-Hong Luo, Jun Xia
ABSTRACT
In the CNS, synapse formation and maturation play crucial roles in the construction and consolidation of neuronal circuits. Neurexin and neuroligin localize on the opposite sides of synaptic membrane and interact with each other to promote the assembly and specialization of synapses. However, the excitatory synapses induced by the neurexin-neuroligin complex are initially immature synapses that lack AMPA receptors. Previously, PICK1 (protein interacting with C kinase 1) was shown to cluster and regulate the synaptic localization of AMPA receptors. Here, we report that during synaptogenesis induced by neurexin in cultured neurons from rat hippocampus, PICK1 recruited AMPA receptors to immature postsynaptic sites. This synaptic recruitment of AMPA receptors depended on the interaction between GluA2 and PICK1, and on the lipid-binding ability of PICK1, but not the interaction between PICK1 and neuroligin. Last, our results demonstrated that the recruitment of GluA2 to synapses could be prevented by ICA69 (islet cell autoantigen 69 kDa), a key binding partner of PICK1. Our study showed that PICK1, being negatively regulated by ICA69, could facilitate synapse maturation.
Sigma-Aldrich
Anti-GRIA2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
Anti-GRIA2 (AB2) antibody produced in rabbit, IgG fraction of antiserum