Merck
  • ROS-dependent Syk and Pyk2-mediated STAT1 activation is required for 15(S)-hydroxyeicosatetraenoic acid-induced CD36 expression and foam cell formation.
ROS-dependent Syk and Pyk2-mediated STAT1 activation is required for 15(S)-hydroxyeicosatetraenoic acid-induced CD36 expression and foam cell formation.
Free radical biology & medicine (2014-08-26)
Sivareddy Kotla, Nikhlesh K Singh, James G Traylor, A Wayne Orr, Gadiparthi N Rao
ABSTRACT
15(S)-Hydroxyeicosatetraenoic acid (15(S)-HETE), the major 15-lipoxygenase 1/2 (15-LO1/2) metabolite of arachidonic acid (AA), induces CD36 expression through xanthine oxidase and NADPH oxidase-dependent ROS production and Syk and Pyk2-dependent STAT1 activation. In line with these observations, 15(S)-HETE also induced foam cell formation involving ROS, Syk, Pyk2, and STAT1-mediated CD36 expression. In addition, peritoneal macrophages from Western diet-fed ApoE(-/-) mice exhibited elevated levels of xanthine oxidase and NADPH oxidase activities, ROS production, Syk, Pyk2, and STAT1 phosphorylation, and CD36 expression compared to those from ApoE(-/-):12/15-LO(-/-) mice and these events correlated with increased lipid deposits, macrophage content, and lesion progression in the aortic roots. Human atherosclerotic arteries also showed increased 15-LO1 expression, STAT1 phosphorylation, and CD36 levels as compared to normal arteries. Together, these findings suggest that 12/15-LO metabolites of AA, particularly 12/15(S)-HETE, might play a crucial role in atherogenesis by enhancing foam cell formation.
Sigma-Aldrich
12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥95% (HPLC)
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oil Red EGN, powder
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, ≥99%
Allopurinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
AG 490, A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC50 = 100 nM).