Merck
  • Ouabain elicits human glioblastoma cells apoptosis by generating reactive oxygen species in ERK-p66SHC-dependent pathway.
Ouabain elicits human glioblastoma cells apoptosis by generating reactive oxygen species in ERK-p66SHC-dependent pathway.
Molecular and cellular biochemistry (2014-09-14)
Xiaofei Yan, FenLi Liang, Dongmin Li, Jin Zheng
ABSTRACT
Excessive reactive oxygen species (ROS) generation has been implicated as one of main agents in ouabain-induced anticancer effect. Unfortunately, the signaling pathways under it are not very clarified. In the present study, we investigated the molecular mechanism involved in ouabain-induced ROS generation and cell apoptosis on human U373MG and U87MG glioma cells. Ouabain-induced glioblastoma cells apoptosis and increased ROS generation. Clearance ROS by three different ROS scavenger partly, but not totally, reversed ouabain's effect on cell apoptosis. Ouabain-induced ROS generation was not regulated by calcium overload, reduced nicotinamide adenine dinucleotide phosphate oxidation, but by p66Shc phosphorylation. Ouabain treatment increased p66Shc Ser36 phosphorylation. Knockdown of p66Shc by siRNA significantly inhibited ROS generations in response to ouabain. Ouabain-induced p66Shc phosphorylation through Src/Ras/extracellular signal-regulated kinase signal pathway. Our results uncovered a novel signaling pathway with p66Shc, ouabain-induced ROS generation, and glioblastoma cell apoptosis.
Sigma-Aldrich
Myxothiazol, from Myxococcus fulvus Mx f85, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cltc
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hc
Sigma-Aldrich
MISSION® esiRNA, targeting human CLTC
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
KB-R7943, ≥98% (HPLC), powder
Sigma-Aldrich
Glutathione Assay Kit, sufficient for 700 assays
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting human C5
Sigma-Aldrich
GW5074, powder
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Ouabain, European Pharmacopoeia (EP) Reference Standard