Merck
  • R-loops associated with triplet repeat expansions promote gene silencing in Friedreich ataxia and fragile X syndrome.
R-loops associated with triplet repeat expansions promote gene silencing in Friedreich ataxia and fragile X syndrome.
PLoS genetics (2014-05-03)
Matthias Groh, Michele M P Lufino, Richard Wade-Martins, Natalia Gromak
ABSTRACT
Friedreich ataxia (FRDA) and Fragile X syndrome (FXS) are among 40 diseases associated with expansion of repeated sequences (TREDs). Although their molecular pathology is not well understood, formation of repressive chromatin and unusual DNA structures over repeat regions were proposed to play a role. Our study now shows that RNA/DNA hybrids (R-loops) form in patient cells on expanded repeats of endogenous FXN and FMR1 genes, associated with FRDA and FXS. These transcription-dependent R-loops are stable, co-localise with repressive H3K9me2 chromatin mark and impede RNA Polymerase II transcription in patient cells. We investigated the interplay between repressive chromatin marks and R-loops on the FXN gene. We show that decrease in repressive H3K9me2 chromatin mark has no effect on R-loop levels. Importantly, increasing R-loop levels by treatment with DNA topoisomerase inhibitor camptothecin leads to up-regulation of repressive chromatin marks, resulting in FXN transcriptional silencing. This provides a direct molecular link between R-loops and the pathology of TREDs, suggesting that R-loops act as an initial trigger to promote FXN and FMR1 silencing. Thus R-loops represent a common feature of nucleotide expansion disorders and provide a new target for therapeutic interventions.
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human RNASEH1
Pricing and availability is not currently available.
Sigma-Aldrich
BIX 01294 trihydrochloride hydrate, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rnaseh1
Pricing and availability is not currently available.
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Pricing and availability is not currently available.