Merck
  • Candida albicans cis-prenyltransferase Rer2 is required for protein glycosylation, cell wall integrity and hypha formation.
Candida albicans cis-prenyltransferase Rer2 is required for protein glycosylation, cell wall integrity and hypha formation.
Fungal genetics and biology : FG & B (2014-05-31)
Mateusz Juchimiuk, Jacek Orłowski, Katarzyna Gawarecka, Ewa Świeżewska, Joachim F Ernst, Grażyna Palamarczyk
ABSTRACT
cis-Prenyltransferase is the first enzyme of the mevalonate pathway committed to the biosynthesis of dolichol in eukaryotes. The RER2 gene encoding cis-prenyltransferase (Rer2p) in the human fungal pathogen Candida albicans was characterized. In addition, the ORF19.5236 encoding the second cis-prenyltransferase, which putatively is responsible for the synthesis of longer polyisoprenoids chains, was identified. When cultivated under repressive conditions, the conditional mutant strain expressing the RER2 gene from the regulatable MET3 promoter contained only 4% of cis-prenyltransferase activity and markedly diminished amounts of dolichols, as compared to the wild-type strain. Moreover, transcriptomal analyses revealed changes in the expression of 300 genes, mainly involved in transport, response to stress, filamentous growth and organelle organization. Growth of the conditional strain was blocked completely at 37 °C. The strain was hypersensitive to a wide range of inhibitors, which suggested glycosylation defects and compromised cell wall integrity. Moreover, the rer2 conditional mutant grown in the repressive conditions, unlike the same strain in the absence of repressor, failed to form hyphae. The results indicate that dolichols are essential not only for protein glycosylation and cell wall integrity but also for growth and development of C. albicans.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Glycerol, analytical standard