Merck
  • Oral iron supplementation after blood donation: a randomized clinical trial.
Oral iron supplementation after blood donation: a randomized clinical trial.
JAMA (2015-02-11)
Joseph E Kiss, Donald Brambilla, Simone A Glynn, Alan E Mast, Bryan R Spencer, Mars Stone, Steven H Kleinman, Ritchard G Cable
ABSTRACT
Although blood donation is allowed every 8 weeks in the United States, recovery of hemoglobin to the currently accepted standard (12.5 g/dL) is frequently delayed, and some donors become anemic. To determine the effect of oral iron supplementation on hemoglobin recovery time (days to recovery of 80% of hemoglobin removed) and recovery of iron stores in iron-depleted ("low ferritin," ≤26 ng/mL) and iron-replete ("higher ferritin," >26 ng/mL) blood donors. Randomized, nonblinded clinical trial of blood donors stratified by ferritin level, sex, and age conducted in 4 regional blood centers in the United States in 2012. Included were 215 eligible participants aged 18 to 79 years who had not donated whole blood or red blood cells within 4 months. One tablet of ferrous gluconate (37.5 mg of elemental iron) daily or no iron for 24 weeks (168 days) after donating a unit of whole blood (500 mL). Time to recovery of 80% of the postdonation decrease in hemoglobin and recovery of ferritin level to baseline as a measure of iron stores. The mean baseline hemoglobin levels were comparable in the iron and no-iron groups and declined from a mean (SD) of 13.4 (1.1) g/dL to 12.0 (1.2) g/dL after donation in the low-ferritin group and from 14.2 (1.1) g/dL to 12.9 (1.2) g/dL in the higher-ferritin group. Compared with participants who did not receive iron supplementation, those who received iron supplementation had shortened time to 80% hemoglobin recovery in both the low-ferritin (mean, 32 days, interquartile range [IQR], 30-34, vs 158 days, IQR, 126->168) and higher-ferritin groups (31 days, IQR, 29-33, vs 78 days, IQR, 66-95). Median time to recovery to baseline ferritin levels in the low-ferritin group taking iron was 21 days (IQR, 12-84). For participants not taking iron, recovery to baseline was longer than 168 days (IQR, 128->168). Median time to recovery to baseline in the higher-ferritin group taking iron was 107 days (IQR, 75-141), and for participants not taking iron, recovery to baseline was longer than 168 days (IQR, >168->168). Recovery of iron stores in all participants who received supplements took a median of 76 days (IQR, 20-126); for participants not taking iron, median recovery time was longer than 168 days (IQR, 147->168 days; P < .001). Without iron supplements, 67% of participants did not recover iron stores by 168 days. Among blood donors with normal hemoglobin levels, low-dose iron supplementation, compared with no supplementation, reduced time to 80% recovery of the postdonation decrease in hemoglobin concentration in donors with low ferritin (≤26 ng/mL) or higher ferritin (>26 ng/mL). clinicaltrials.gov Identifier: NCT01555060.
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Iron, foil, light tested, 50x50mm, thickness 0.05mm, hard, 99.5%
Iron, foil, not light tested, 25x25mm, thickness 0.009mm, 99.85%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 8mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, not light tested, 25x25mm, thickness 0.002mm, 99.85%
Iron, foil, 8mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, not light tested, 300x300mm, thickness 0.05mm, hard, 99.5%
Iron, rod, 300mm, diameter 5.0mm, as drawn, 99.99+%
Iron, foil, light tested, 25x25mm, thickness 0.025mm, hard, 99.5%
Iron, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.006mm, 99.85%
Iron, foil, not light tested, 150x150mm, thickness 0.008mm, 99.85%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.0125mm, 99.85%
Iron, foil, 1m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.5mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 20m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.008mm, 99.85%