Merck
  • Attenuation of inflammatory response by 25- hydroxyvitamin D3-loaded polylactic acid microspheres in treatment of periodontitis in diabetic rats.
Attenuation of inflammatory response by 25- hydroxyvitamin D3-loaded polylactic acid microspheres in treatment of periodontitis in diabetic rats.
The Chinese journal of dental research : the official journal of the Scientific Section of the Chinese Stomatological Association (CSA) (2014-12-23)
Hao Li, Bo Li, Qi Wang, Yu Xiao, Xin Min Chen, Wei Li
ABSTRACT
To investigate the inhibitory effects of 25-hydroxyvitamin D3 (25(OH)D3)-loaded polylactic acid (PLA) microspheres on inflammatory response in diabetic periodontitis. 25(OH)D3-loaded PLA microspheres were produced using emulsion-solvent evaporation method. Rat bone marrow stromal cells (BMSCs) cultured with Aggregatibacter actinomycetemcomitans in high glucose medium were chosen to mimic diabetic periodontitis. After cultivation with 25(OH)D3-loaded PLA microspheres, inflammation-related proteins in BMSCs were detected using Western blot analysis. Periodontitis was induced using silk ligatures in diabetic rats, and 25(OH)D3-loaded PLA microspheres were placed into the periodontal pockets. Periodontal tissues were examined using hematoxylin and eosin staining and western blot analysis. Drug release from the 25(OH)D3-loaded microspheres was relatively steady during 70 days. In a diabetic periodontitis-like environment, 25(OH)D3-loaded microspheres upregulated vitamin D receptor expression, and downregulated nuclear factor-κB expression and signal transducer and activator of transcription 3 phosphorylation in the BMSCs. These 25(OH)D3 microspheres also attenuated periodontal inflammatory infiltrate and bone loss in diabetic rats with periodontitis. 25(OH)D3-loaded microspheres could ameliorate diabetic periodontitis by inhibiting inflammatory response, and may provide a potential therapy for patients with this disease.
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Polylactic acid, Mw ~60,000
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%