Merck
  • Self-adhesive resin cements: adhesive performance to indirect restorative ceramics.
Self-adhesive resin cements: adhesive performance to indirect restorative ceramics.
The journal of adhesive dentistry (2014-12-18)
José Zorzin, Renan Belli, Andrea Wagner, Anselm Petschelt, Ulrich Lohbauer
ABSTRACT
To evaluate the bonding performance of self-adhesive resin cements to zirconia and lithium disilicate in self- and dual-curing modes before and after thermocycling. Rectangular bars (3 mm high, 3 mm wide, 9 mm long) were manufactured from zirconia (Vita In-Ceram YZ for inLab, VITA) and lithium disilicate blocks (IPS e.max Press, Ivoclar Vivadent) (n=240 per material). Zirconia bars were sandblasted (35 μm Al2O3, 1.5 bar pressure). Lithium disilicate bars were HF etched (20 s, IPS Ceramic Etching Gel, Ivoclar Vivadent) and silanized with ESPE Sil (3M ESPE). Forty bars of zirconia were luted in twos perpendicular to each other as were 40 bars of lithium disilicate using RelyX Unicem Automix 2 (3M ESPE), G-Cem LinkAce (GC Europe) or Maxcem Elite (Kerr) in self- or dual-curing mode. Half of the specimens from each material were submitted to tensile bond strength (TBS) testing after 24-h storage in distilled water at 37°C, and half underwent TBS testing after thermocycling (5000 cycles, 5°C/55°C, 30-s dwell time). Bond strength values for each bonding substrate were analyzed using one-way ANOVA (Student-Newman- Keuls, α=0.05). On zirconia, dual-curing resulted in significantly (p<0.05) higher tensile bond strengths compared to self-curing, with the exception of RelyX Unicem 2 after thermocycling. Thermocycling significantly (p<0.05) reduced the tensile bond strength of Maxcem Elite to zirconia in both curing modes. The TBS of self-adhesive cements to lithium disilicate showed no significant (p>0.05) difference between the different curing modes and after thermocycling. For most of the investigated self-adhesive cements, bond strengths to zirconia were increased by dual curing; this was not true for lithium disilicate. For luting on zirconia with self-adhesive cements, dual curing is strongly recommended in clinical situations.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 100x100mm, thickness 0.25mm, annealed, 99.2%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Zirconium, foil, not light tested, 25x25mm, thickness 0.003mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.015mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.006mm, 99.2%
Zirconium, foil, 8mm disks, thickness 0.25mm, annealed, 99.2%