Merck
  • Bond strength and monomer conversion of indirect composite resin restorations, Part 1: Light vs heat polymerization.
Bond strength and monomer conversion of indirect composite resin restorations, Part 1: Light vs heat polymerization.
The journal of adhesive dentistry (2014-12-18)
Daniel Alexandre Menezes Pedrosa Malta, Pascal Magne, Sylvio Monteiro-Junior
ABSTRACT
To assess the resin microtensile bond strength (MTBS) and the monomer conversion (MC) of indirect composite resin restorations made of three different materials. Two light-polymerized direct materials (Filtek Z100 and Premise) and one light- and heat-polymerized indirect material (Premise Indirect) were used. For MTBS testing, 42 cylindrical samples were fabricated (7 pairs per material). Surface conditioning included airborne-particle abrasion, cleaning, and application of a silane. Cylinders were bonded to each other using adhesive resin (Optibond FL). Specimens were stored in water for 24 h. Another 15 cylinders (5 per material) were fabricated for MC measurements (FT-IR) immediately and at 24 h. The MTBS data were submitted to one-way ANOVA and the MC to two-way ANOVA (material and storage time) (α=0.05), followed by post-hoc comparisons with the Tukey test. The MTBS to Z100 was 72.2 MPa, significantly higher than that to Premise (48.4 MPa) and Premise Indirect (52.7 MPa). The immediate MC was similar for all materials (range 51% to 56%) and significantly increased at 24 h (range 57% to 66%), except for Z100. Premise Indirect showed the highest MC (66% at 24 h). Z100 showed better "bondability" than Premise and Premise Indirect. Premise Indirect, with its heat initiator, did not present a higher MC.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 100x100mm, thickness 0.25mm, annealed, 99.2%
Sigma-Aldrich
Water, for molecular biology, sterile filtered