Merck
  • Adjuvant granulocyte colony-stimulating factor therapy results in improved spatial learning and stimulates hippocampal neurogenesis in a mouse model of pneumococcal meningitis.
Adjuvant granulocyte colony-stimulating factor therapy results in improved spatial learning and stimulates hippocampal neurogenesis in a mouse model of pneumococcal meningitis.
Journal of neuropathology and experimental neurology (2014-12-04)
Anna Kathrin Schmidt, Arno Reich, Björn Falkenburger, Jörg B Schulz, Lars Ove Brandenburg, Sandra Ribes, Simone C Tauber
ABSTRACT
Despite the development of new antibiotic agents, mortality of pneumococcal meningitis remains high. In addition, meningitis results in severe long-term morbidity, most prominently cognitive deficits. Granulocyte colony-stimulating factor (G-CSF) stimulates proliferation and differentiation of hematopoietic progenitor cells and increases the number of circulating neutrophil granulocytes. This study investigated the effect of adjuvant G-CSF treatment on cognitive function after pneumococcal meningitis. C57BL/6 mice were infected by subarachnoid injection of Streptococcus pneumoniae serotype 3 and treated with ceftriaxone and G-CSF subcutaneously or ceftriaxone alone for 5 days. Clinical scores, motor performance, and mortality during bacterial meningitis were unaffected by adjuvant G-CSF treatment. No effect of G-CSF treatment on production of proinflammatory cytokines or activation of microglia or astrocytes was observed. The G-CSF treatment did, however, result in hippocampal neurogenesis and improved spatial learning performance 6 weeks after meningitis. These results suggest that G-CSF might offer a new adjuvant therapeutic approach in bacterial meningitis to reduce long-term cognitive deficits.
Sigma-Aldrich
Xylazine, ≥99%
Supelco
Ceftriaxone Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic
USP
Ceftriaxone sodium, United States Pharmacopeia (USP) Reference Standard
Ceftriaxone sodium, European Pharmacopoeia (EP) Reference Standard