Merck
  • Myricetin prevents titanium particle-induced osteolysis in vivo and inhibits RANKL-induced osteoclastogenesis in vitro.
Myricetin prevents titanium particle-induced osteolysis in vivo and inhibits RANKL-induced osteoclastogenesis in vitro.
Biochemical pharmacology (2014-12-03)
Chuanlong Wu, Wengang Wang, Bo Tian, Xuqiang Liu, Xinhua Qu, Zanjing Zhai, Haowei Li, Fengxiang Liu, Qiming Fan, Tingting Tang, An Qin, Zhenan Zhu
ABSTRACT
Titanium (Ti) particle-induced periprosthetic osteolysis and subsequent aseptic loosening are a primary reason for total hip arthroplasty failure. The aim of this study was to assess the effect of myricetin on Ti particle-induced osteolysis and osteoclastogenesis. We demonstrated that myricetin, a natural plant extract, exerts potent inhibitory effects on Ti particle-induced osteolysis in a mouse calvarial model. Further histological analysis indicated that the inhibition of osteoclast formation and function, and the secretion of inflammatory factors, are key targets for therapeutic agents in the treatment of wear particle-induced osteolysis. In vitro, we found that myricetin suppressed receptor activator of nuclear factor-κB ligand (RANKL)-mediated osteoclast differentiation, bone resorption, and F-actin ring formation in a dose-dependent manner. Moreover, myricetin significantly reduced the expression of osteoclast-specific markers in mouse bone marrow-derived macrophages, including tartrate-resistant acid phosphatase (TRAP), cathepsin K, the calcitonin receptor, V-ATPase d2, c-fos, and nuclear factor of activated T cells (NFAT) c1. Further investigation revealed that myricetin inhibited osteoclastogenesis through the suppression of the nuclear factor-κB (NF-κB) signaling pathway and mitogen-activated protein kinase (MAPK) pathways involving extracellular signal-regulated kinase 1/2 (ERK1/2), p38, and c-Jun N-terminal kinase 1/2 (JNK1/2). While, the inhibition of TNF-α and IL-1β secretion was another reason for the suppressive effect of myricetin on Ti particle-induced osteolysis. Collectively, these findings suggest that myricetin is a potential natural agent for the treatment of periprosthetic osteolysis and other osteoclast-related osteolytic diseases.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Glycine
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis