Merck
  • Concentrations of phthalate metabolites in breast milk in Korea: estimating exposure to phthalates and potential risks among breast-fed infants.
Concentrations of phthalate metabolites in breast milk in Korea: estimating exposure to phthalates and potential risks among breast-fed infants.
The Science of the total environment (2014-12-02)
Sunmi Kim, Jangwoo Lee, Jeongim Park, Hai-Joong Kim, Geumjoon Cho, Gun-Ha Kim, So-Hee Eun, Jeong Jae Lee, Gyuyeon Choi, Eunsook Suh, Sooran Choi, Sungjoo Kim, Young Don Kim, Sung Koo Kim, Su Young Kim, Seunghyo Kim, Soyong Eom, Hyo-Bang Moon, Sungkyoon Kim, Kyungho Choi
ABSTRACT
Phthalates have been associated with endocrine disruption and developmental effects in many experimental and epidemiological studies. Developing infants are among the most susceptible populations to endocrine disruption. However, limited information is available on phthalate exposure and its associated risks among breast-fed newborn infants. In the present study, breast milk samples were collected from 62 lactating mothers at 1 month post-partum from four cities of Korea in 2012 and were evaluated for six phthalate metabolites (mono-isobutyl phthalate (MiBP), mono-n-butyl phthalate (MnBP), mono(2-ethyl-hexyl) phthalate (MEHP), mono-(2-ethyl-5-hydroxyhexyl) phthalate (MEHHP), mono-(2-ethyl-5-oxohexyl) phthalate (MEOHP) and monoethyl phthalate (MEP)). MEP was detected in all breast milk samples, with a median concentration of 0.37 μg/L, and MiBP, MnBP and MEHP were detected in 79-89% of samples, with median concentrations of 1.10, 1.70, and 2.08 μg/L, respectively. However, MEHHP and MEOHP, the oxidized forms of di-ethyl-hexyl phthalate (DEHP), were detected in only one sample. For exposure assessment, the levels of phthalate diesters were estimated based on the parent:metabolite ratios in the breast milk that are reported elsewhere. For risk assessment, the endocrine-related toxicity of the monoester was assumed to be the same as that of its diester form. Median daily intake estimates of phthalates, including both monoester and diester forms, through breast milk consumption ranged between 0.91 and 6.52 μg/kg body weight (bw) for DEHP and between 0.38 and 1.43 μg/kg bw for di-n-butyl phthalate (DnBP). Based on the estimated daily intake, up to 8% of infants exceeded the reference dose of anti-androgenicity (RfD AA) for DEHP, and 6% of infants exceeded the tolerable daily intake (TDI) for DnBP. Breast milk MiBP and MnBP concentrations showed significant positive associations with maternal consumption of whipped cream or purified water. Considering vulnerability of young infants, efforts to mitigate phthalate exposure among lactating women are warranted.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Ammonium acetate, Vetec, reagent grade, 97%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard