Merck
  • Tissue-specific splice variants of HARE/Stabilin-2 are expressed in bone marrow, lymph node, and spleen.
Tissue-specific splice variants of HARE/Stabilin-2 are expressed in bone marrow, lymph node, and spleen.
Biochemical and biophysical research communications (2014-12-03)
Amanda K Hare, Edward N Harris
ABSTRACT
The hyaluronan receptor for endocytosis (HARE), or Stabilin-2, is the mammalian endocytic clearance receptor for HA, heparin, advanced glycation end-products, acetylated and oxidized low-density lipoproteins and collagen N-terminal propeptides. This large 2551 amino acid receptor is encoded by a gene that covers over 180 kbp on human chromosome 12 and is predicted to be composed of 69 exons. Due to the expression profile of this gene and the number of exons it contains, we hypothesized that splice variants of stab2 are encoded in these tissues. In addition, a correlation between alternative splice variants and cancer progression has been shown in other HA receptors such as RHAMM and CD42. In this study, two methods were utilized in identifying and/or isolating the HARE splice variants. The first method used primer sets to amplify the 190-HARE encoding region that could contain splice junctions; therefore, they were purified from agarose gels and sequenced. Five splice variants were detected in that manner. In the second approach, the entire open reading frame of HARE was amplified. This allowed four splice variants with extensive exon splicing to be isolated. After the splice variants were sequenced, three were cloned into a mammalian expression vector. Next, stable cell lines expressing the variants were created in order to determine stable protein expression. In this study, the splice variants were found to be tissue specific in most cases. This suggests that tissue specific regulatory splicing mechanisms may lead to differences in functionality between the splice variants.
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance