Merck
  • Young patients with non-germinal center B-cell-like diffuse large B-cell lymphoma benefit from intensified chemotherapy with ACVBP plus rituximab compared with CHOP plus rituximab: analysis of data from the Groupe d'Etudes des Lymphomes de l'Adulte/lymphoma study association phase III trial LNH 03-2B.
Young patients with non-germinal center B-cell-like diffuse large B-cell lymphoma benefit from intensified chemotherapy with ACVBP plus rituximab compared with CHOP plus rituximab: analysis of data from the Groupe d'Etudes des Lymphomes de l'Adulte/lymphoma study association phase III trial LNH 03-2B.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-11-12)
Thierry Jo Molina, Danielle Canioni, Christiane Copie-Bergman, Christian Recher, Josette Brière, Corinne Haioun, Françoise Berger, Christophe Fermé, Marie-Christine Copin, Olivier Casasnovas, Catherine Thieblemont, Tony Petrella, Karen Leroy, Gilles Salles, Bettina Fabiani, Franck Morschauser, Nicolas Mounier, Bertrand Coiffier, Fabrice Jardin, Philippe Gaulard, Jean-Philippe Jais, Hervé Tilly
ABSTRACT
To determine whether any tumor biomarkers could account for the survival advantage observed in the LNH 03-2B trial among patients with diffuse large B-cell lymphoma (DLBCL) and low-intermediate risk according to the International Prognostic Index when treated with dose-intensive rituximab, doxorubicin, cyclophosphamide, vindesine, bleomycin, and prednisone (R-ACVBP) compared with standard rituximab, doxorubicin, cyclophosphamide, vincristine, and prednisone (R-CHOP). Using immunohistochemistry, expression of CD10, BCL6, MUM1, MYC, and BCL2 and coexpression of MYC/BCL2 were examined. The interaction effects between each biomarker and treatment arm on survival were studied in a restricted model and a full model incorporating clinical parameters. Among the 379 patients analyzed in the trial, 229 tumors were evaluable for germinal center B-cell-like (GCB)/non-GCB subclassification according to the Hans algorithm. Among all the biomarkers, only the interaction between the Hans algorithm and the treatment arm was significant for progression-free survival (PFS) and overall survival (OS) in univariable (PFS, P = .04; OS, P = .01) and multivariable (PFS, P = .03; OS, P = .01) analyses. Non-GCB tumors predicted worse PFS (hazard ratio [HR], 3.21; 95% CI, 1.29 to 8.00; P = .01) and OS (HR, 6.09; 95% CI, 1.37 to 27.03; P = .02) among patients treated with R-CHOP compared with patients who received R-ACVBP, whereas there were no significant survival differences between these regimens among patients with GCB tumors. The survival benefit related to R-ACVBP over R-CHOP is at least partly linked to improved survival among patients with non-GCB DLBCL. Therefore, the Hans algorithm could be considered a theragnostic biomarker for selecting young patients with DLBCL who can benefit from an intensified R-ACVBP immunochemotherapy regimen.
Product Description
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vindesine sulfate salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Prednisolone, ≥99%
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Supelco
Prednisolone, VETRANAL®, analytical standard
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Vindesine sulfate, European Pharmacopoeia (EP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard