Merck
  • Crystal structure of a DNA/Ba2+ G-quadruplex containing a water-mediated C-tetrad.
Crystal structure of a DNA/Ba2+ G-quadruplex containing a water-mediated C-tetrad.
Nucleic acids research (2014-11-13)
Diana Zhang, Terry Huang, Philip S Lukeman, Paul J Paukstelis
ABSTRACT
We have determined the 1.50 Å crystal structure of the DNA decamer, d(CCA(CNV)KGCGTGG) ((CNV)K, 3-cyanovinylcarbazole), which forms a G-quadruplex structure in the presence of Ba(2+). The structure contains several unique features including a bulged nucleotide and the first crystal structure observation of a C-tetrad. The structure reveals that water molecules mediate contacts between the divalent cations and the C-tetrad, allowing Ba(2+) ions to occupy adjacent steps in the central ion channel. One ordered Mg(2+) facilitates 3'-3' stacking of two quadruplexes in the asymmetric unit, while the bulged nucleotide mediates crystal contacts. Despite the high diffraction limit, the first four nucleotides including the (CNV)K nucleoside are disordered though they are still involved in crystal packing. This work suggests that the bulky hydrophobic groups may locally influence the formation of non-Watson-Crick structures from otherwise complementary sequences. These observations lead to the intriguing possibility that certain types of DNA damage may act as modulators of G-quadruplex formation.
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride dihydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis