Merck
  • Evaluation of acidity estimation methods for mine drainage, Pennsylvania, USA.
Evaluation of acidity estimation methods for mine drainage, Pennsylvania, USA.
Environmental monitoring and assessment (2014-11-17)
Daeryong Park, Byungtae Park, Justin J Mendinsky, Benjaphon Paksuchon, Ratda Suhataikul, Brian A Dempsey, Yunchul Cho
ABSTRACT
Eighteen sites impacted by abandoned mine drainage (AMD) in Pennsylvania were sampled and measured for pH, acidity, alkalinity, metal ions, and sulfate. This study compared the accuracy of four acidity calculation methods with measured hot peroxide acidity and identified the most accurate calculation method for each site as a function of pH and sulfate concentration. Method E1 was the sum of proton and acidity based on total metal concentrations; method E2 added alkalinity; method E3 also accounted for aluminum speciation and temperature effects; and method E4 accounted for sulfate speciation. To evaluate errors between measured and predicted acidity, the Nash-Sutcliffe efficiency (NSE), the coefficient of determination (R (2)), and the root mean square error to standard deviation ratio (RSR) methods were applied. The error evaluation results show that E1, E2, E3, and E4 sites were most accurate at 0, 9, 4, and 5 of the sites, respectively. Sites where E2 was most accurate had pH greater than 4.0 and less than 400 mg/L of sulfate. Sites where E3 was most accurate had pH greater than 4.0 and sulfate greater than 400 mg/L with two exceptions. Sites where E4 was most accurate had pH less than 4.0 and more than 400 mg/L sulfate with one exception. The results indicate that acidity in AMD-affected streams can be accurately predicted by using pH, alkalinity, sulfate, Fe(II), Mn(II), and Al(III) concentrations in one or more of the identified equations, and that the appropriate equation for prediction can be selected based on pH and sulfate concentration.
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 6.0mm, as drawn, 99.999%
Aluminum, microfoil, disks, 50mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, rod, 44mm, diameter 77.5mm, as drawn, 99.999%
Aluminum, tube, 200mm, outside diameter 0.86mm, inside diameter 0.76mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, microfoil, disks, 25mm, thinness 0.0075μm, specific density 1.84μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, wire reel, 100m, diameter 0.5mm, as drawn, 99.95+%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, tube, 100mm, outside diameter 0.6mm, inside diameter 0.3mm, wall thickness 0.15mm, as drawn, 99.5%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, tube, 200mm, outside diameter 1.42mm, inside diameter 1.166mm, wall thickness 0.127mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 10.0mm, as drawn, 99.999%
Aluminum, tube, 500mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, sphere, 200pcs, diameter 6.0mm, 99%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, wire reel, 100m, diameter 0.025mm, as drawn, 99.0+%
Aluminum, rod, 200mm, diameter 12.7mm, 99%