Merck
Iron, inflammation, and early death in adults with sickle cell disease.
Circulation research (2014-11-08)
Eduard J van Beers, Yanqin Yang, Nalini Raghavachari, Xin Tian, Darlene T Allen, James S Nichols, Laurel Mendelsohn, Sergei Nekhai, Victor R Gordeuk, James G Taylor, Gregory J Kato
ABSTRACT
Patients with sickle cell disease (SCD) have markers of chronic inflammation, but the mechanism of inflammation and its relevance to patient survival are unknown. To assess the relationship between iron, inflammation, and early death in SCD. Using peripheral blood mononuclear cell transcriptome profile hierarchical clustering, we classified 24 patients and 10 controls in clusters with significantly different expression of genes known to be regulated by iron. Subsequent gene set enrichment analysis showed that many genes associated with the high iron cluster were involved in the toll-like receptor system (TLR4, TLR7, and TLR8) and inflammasome complex pathway (NLRP3, NLRC4, and CASP1). Quantitative PCR confirmed this classification and showed that ferritin light chain, TLR4, and interleukin-6 expression were >100-fold higher in patients than in controls (P<0.001). Further linking intracellular iron and inflammation, 14 SCD patients with a ferroportin Q248H variant that causes intracellular iron accumulation had significantly higher levels of interleukin-6 and C-reactive protein compared with 14 matched SCD patients with the wild-type allele (P<0.05). Finally, in a cohort of 412 patients followed for a median period of 47 months (interquartile range, 24-82), C-reactive protein was strongly and independently associated with early death (hazard ratio, 3.0; 95% confidence interval, 1.7-5.2; P<0.001). Gene expression markers of high intracellular iron in patients with SCD are associated with markers of inflammation and mortality. The results support a model in which intracellular iron promotes inflammatory pathways, such as the TLR system and the inflammasome, identifying important new pathways for additional investigation.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
Iron, foil, 300x300mm, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Iron, wire reel, 1000m, diameter 0.125mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.005mm, 99.85%
Iron, tube, 500mm, outside diameter 3.18mm, inside diameter 1.96mm, wall thickness 0.61mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Iron, wire reel, 20m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1000m, diameter 0.25mm, hard, 99.5%
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%