Merck
  • Th2 cytokine-primed airway smooth muscle cells induce mast cell chemotaxis via secretion of ATP.
Th2 cytokine-primed airway smooth muscle cells induce mast cell chemotaxis via secretion of ATP.
The Journal of asthma : official journal of the Association for the Care of Asthma (2014-10-02)
Ya-Dong Gao, Jie Cao, Ping Li, Ge Huang, Jiong Yang
ABSTRACT
Mast cell infiltration into airway smooth muscle (ASM) bundle is an important feature of asthma. Extracellular adenosine triphosphate (eATP) contributes to the initiation of airway inflammation. eATP induces mast cells migration by acting through purinergic receptors. CD39 is an ectonucleotidase that degrades ATP to ADP and AMP. Whether eATP participates in the migration of mast cell towards ASM cells is still unknown. Airway smooth muscle cells (ASMCs) were isolated from C57/BL6J mice sensitized and challenged with OVA. ASMCs were in vitro cultured and stimulated with IL-4 + IL-13 in the presence or absence of exogenous CD39 or CD39 inhibitor ARL67156. ATP level in the supernatants was measured with ATP content determination kit. CXCL10 concentration in the ASMCs supernatants was measured by ELISA, the mRNA expression of CXCL10 in ASMCs was determined with real-time PCR. Human mast cell line HMC-1 was cultured in Iscove's-Modified Dubecco's Medium. The expression of CXCR3 in HMC-1 cells was determined with flow cytometry and real-time PCR, respectively. HMC-1 migration rates were determined with transwell system. In the supernatants of Th2 cytokine-stimulated ASMCs, ATP level was higher than that without stimulation. CD39 decreased, whereas ARL67156 increased ATP level in the supernatants. Both ATP and the supernatants of Th2 cytokine-stimulated ASMCs induced migration of HMC-1 cells. The surface and mRNA expression of CXCR3 in HMC-1 cells, and the mRNA expression and secretion of CXCL10 in ASMCs were increased after stimulation with ATP or Th2 cytokines. All these effects were partially inhibited by CD39. Our data suggested ASMCs in the asthma microenvironment promoted the migration of mast cells via secretion of ATP and the expression of CXCL10/CXCR3 axis. CD39 could reverse this effect and may be a new target for the treatment of asthma.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Human IP-10 / CXCL10 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse CRG-2 / CXCL10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%