Merck
  • Transformations of citrate and Tween coated silver nanoparticles reacted with Na₂S.
Transformations of citrate and Tween coated silver nanoparticles reacted with Na₂S.
The Science of the total environment (2014-09-30)
M Baalousha, K P Arkill, I Romer, R E Palmer, J R Lead
ABSTRACT
Silver nanoparticles (Ag NPs) are susceptible to transformations in environmental and biological media such as aggregation, oxidation, dissolution, chlorination, sulfidation, formation/replacement of surface coatings following interaction with natural organic matter (NOM). This paper investigates the impact of surface coating and Suwannee River fulvic acid (SRFA) on the transformations and behavior of Ag NPs (citrate coated and Tween coated; cit-Ag NPs and Tween-Ag NPs, respectively), following reaction with different concentrations of Na2S solution (as a source of sulfide species, H2S and HS(-)). These transformations and the dominant mechanisms of transformations were investigated using UV-vis and scanning transmission electron microscopy coupled with electron energy loss spectroscopy. Here, we have shown that Ag NP surface coating impacts their dissolution following dilution in ultrahigh purity water, with higher extent of dissolution of Tween-Ag NPs compared with cit-Ag NPs. Tween-Ag NPs are susceptible to dissolution following their sulfidation at low S/Ag molar ratio. Suwannee River fulvic acid (SRFA) slows down the dissolution of Tween-Ag NPs at low sulfide concentrations and reduces the aggregation of cit-Ag NP in the presence of sodium sulfide. Sulfidation appears to occur by direct interaction of sulfide species with Ag NPs rather than by indirect reaction of sulfide with dissolved Ag species subsequent to dissolution. Furthermore, the sulfidation process results in the formation of partially sulfidized Ag NPs containing unreacted (metallic) subgrains at the edge of the NPs for Tween-Ag NPs in the presence of high sulfide concentration (2000nM Na2S), which occurred to less extent at lower Na2S concentration for Tween-Ag NPs and at all concentrations of Na2S for cit-Ag NPs. Thus, sulfidized Ag NPs may preserve some of the properties of the Ag NPs such as their potential to shed Ag(+) ions and their toxic potential of Ag NPs.
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Sodium sulfide
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Silver, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.004mm, 99.97%
Silver, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, wire reel, 1m, diameter 0.025mm, as drawn, 99.99%
Silver, wire reel, 20m, diameter 0.125mm, annealed, 99.99%
Silver, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95+%
Silver, wire reel, 1m, diameter 0.050mm, gold plated, 99.99%
Silver, wire reel, 0.5m, diameter 0.8mm, as drawn, 99.99%
Silver, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.008mm, 99.95+%
Silver, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 10mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%