Merck
  • Classical against molecular-genetic methods for susceptibility testing of antituberculotics.
Classical against molecular-genetic methods for susceptibility testing of antituberculotics.
Advances in experimental medicine and biology (2014-09-26)
I Porvaznik, J Mokry, I Solovic
ABSTRACT
Tuberculosis currently belongs to rare respiratory diseases in Slovakia. However, the emergence and spread of multi-drug resistant tuberculosis (MDR-TB) and extensively drug-resistant tuberculosis (XDR-TB) are major challenges for global tuberculosis control, since the treatment of resistant forms creates both medical and financial problems. Cultivation methods of diagnosis are time-consuming, many times exceeding the time of the initial phase of tuberculosis treatment. Therefore, in the presented study we compared the standard procedures, based on the cultivation of mycobacteria and subsequent drug susceptibility testing to antituberculotics, with molecular-genetic methods using PCR diagnostic kits. The molecular-genetic testing enables to obtain direct and fast evidence of Mycobacterium tuberculosis, with genomic verification of resistance to the most important anti-tuberculosis drugs - isoniazid and rifampicin in MDR-TB, and ethambutol, aminoglycosides, and fluoroquinolones in XDR-TB. In 2012-2013, we confirmed 19 cases of drug-resistant tuberculosis in Slovakia. The resistance to rifampicin was confirmed in all strains with both methods. In two cases, the molecular-genetic testing did not show resistance to isoniazid, as confirmed by conventional cultivation. Furthermore, two strains demonstrating susceptibility in conventional microbiological testing to ethambutol and five strains to fluoroquinolones were verified as actually being resistant using a PCR method. Rapid diagnosis and identification of MDR-TB or XDR-TB strains using molecular-genetic testing is an essential tool for the timely and appropriate drug treatment and prevention of spread of drug resistant strains.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ethambutol dihydrochloride, antimycobacterial
Ethambutol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Ethambutol for system suitability, European Pharmacopoeia (EP) Reference Standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Supelco
Isoniazid, Vetec, reagent grade, 98%
Rifampicin, European Pharmacopoeia (EP) Reference Standard