Merck
  • Mycophenolic acid in the treatment of birdshot chorioretinopathy: long-term follow-up.
Mycophenolic acid in the treatment of birdshot chorioretinopathy: long-term follow-up.
The British journal of ophthalmology (2014-07-26)
Deshka Doycheva, Herbert Jägle, Manfred Zierhut, Christoph Deuter, Gunnar Blumenstock, Ulrich Schiefer, Katarina Stingl, Kai Januschowski, Bogomil Voykov, Nicole Stuebiger
ABSTRACT
To assess the long-term efficacy and tolerability of both derivatives of mycophenolic acid, mycophenolate mofetil (MMF) and mycophenolate sodium (MPS), in the therapy of patients with birdshot chorioretinopathy (BSCR). Retrospective analysis of 24 patients (48 eyes) with BSCR, treated with MMF or MPS with a follow-up of at least 1 year. The main outcome measures included control of inflammation, steroid-sparing potential and side effects. Secondary outcome measure was the development of retinal function during the therapy measured by best-corrected visual acuity (BCVA), visual field and/or electroretinography (ERG). Twelve patients (50%) were treated with MMF and 12 patients (50%) with MPS. Control of intraocular inflammation, defined as complete lack of clinical and angiographic signs of inflammatory activity, was achieved in 16 of 24 patients (67%). The angiographic signs of activity were significantly reduced during the follow-up (p<0.05). No significant difference was found in the mean BCVA, the visual field and the ERG parameters during the treatment compared with the baseline (p>0.05). In 20 out of 21 patients (95%) who received systemic corticosteroids, the corticosteroids could be tapered to a daily dose of ≤10 mg (rate 0.26/patient-year). Drug-related side effects occurred in 12 patients (50%, rate 0.16/patient-year). In four patients (17%), a therapy switch from MMF to MPS was undertaken due to gastrointestinal discomfort. Derivatives of mycophenolic acid are effective and safe drugs for the treatment of BSCR. In cases with gastrointestinal side effects, a therapy switch from MMF to MPS should be considered.
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
USP
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Mycophenolic acid, analytical standard