Merck
  • Immunophenotypic profile of myeloid cells in granulocytic sarcoma by immunohistochemistry. Correlation with blast differentiation in bone marrow.
Immunophenotypic profile of myeloid cells in granulocytic sarcoma by immunohistochemistry. Correlation with blast differentiation in bone marrow.
American journal of clinical pathology (2000-11-09)
C C Chang, C Eshoa, B Kampalath, V B Shidham, S Perkins
ABSTRACT
The present study was designed to evaluate the lineage differentiation (particularly monocytic differentiation) of immature myeloid cells in granulocytic sarcoma (GS) by immunohistochemistry and correlate the results with lineage differentiation of blasts in the bone marrow and to determine the degree of maturation of the infiltrating myeloid cells in GS by immunohistochemistry using CD34 and HLA-DR. Immunohistochemical stains were performed on paraffin-embedded tissue from 17 GS lesions with lineage-associated markers: myeloperoxidase, CD68 (KP1), CD68 (PG-Ml), glycophorin A, factor VIII, and CD56; and with markers for blasts and immature myeloid cells: CD34 and HLA-DR. Our results show that positive staining with PG-M1, but not KP1, suggests monocytic differentiation of myeloid cells in GS and correlates with the monocytic differentiation of blasts in the bone marrow. Expression of CD56 is frequent in GS, especially when the marrow blasts have monocytic differentiation, and should not be interpreted as a primary natural-killer cell process. The immature myeloid cells in GS are frequently HLA-DR positive. However, CD34 positivity of the immature myeloid cells is relatively uncommon, except in cases with underlying myelodysplastic syndrome or chronic myelogenous leukemia.