Merck
  • Kinetin inhibits apoptosis of aging spleen cells induced by D-galactose in rats.
Kinetin inhibits apoptosis of aging spleen cells induced by D-galactose in rats.
Journal of veterinary science (2014-06-26)
Mengyun Li, Wuqing Ouyang, Xiaoli Wu, Yin Zheng, Yunpeng Wei, Lei An
ABSTRACT
Kinetin (Kn) is a cytokinin growth factor that exerts several anti-aging and antioxidant effects on cells and organs. To investigate the mechanism underlying apoptotic events in aging cells induced by D-galactose (D-gal), we examined the effect of Kn delivered via nuchal subcutaneous injection on D-gal-induced aging and apoptosis in rats. Our results showed that interleukin (IL)-2 levels and mitochondrial membrane potential (ΔΨm) were decreased by Kn in aging rats while IL-6 production and apoptosis increased. In addition, the expression of anti-apoptotic Bcl-2 was low while that of Bax was high in the aging group. After treated with Kn, compared with aging group, there showed obvious difference in Kn group with elevated IL-2, proliferation index, Bcl-2, ΔΨm and decreased IL-6 and Bax in splenic lymphocyte. Based on these results, we concluded that Kn can effectively protect the rat spleen from aging, apoptosis, and atrophy.
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture