Merck
  • Red-emitting rhodamines with hydroxylated, sulfonated, and phosphorylated dye residues and their use in fluorescence nanoscopy.
Red-emitting rhodamines with hydroxylated, sulfonated, and phosphorylated dye residues and their use in fluorescence nanoscopy.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-09-13)
Kirill Kolmakov, Christian A Wurm, René Hennig, Erdmann Rapp, Stefan Jakobs, Vladimir N Belov, Stefan W Hell
ABSTRACT
Fluorescent dyes emitting red light are frequently used in conventional and super-resolution microscopy of biological samples, although the variety of the useful dyes is limited. We describe the synthesis of rhodamine-based fluorescent dyes with absorption and emission maxima in the range of 621-637 and 644-660 nm, respectively and demonstrate their high performance in confocal and stimulated emission depletion (STED) microscopy. New dyes were prepared by means of reliable chemical transformations applied to a rhodamine scaffold with three variable positions. They feature polarity, water solubility, variable net charges, improved stabilities of N-hydroxysuccinimidyl (NHS) esters, as well as large fluorescence quantum yields in dye solutions and antibody conjugates. The photophysical and imaging properties of dyes containing three different polar groups, namely primary phosphate, sulfonic acid (in two different positions), and hydroxyl were compared. A dye with two primary phosphate groups was explored as a valuable alternative to dyes with "classical" sulfonic acid groups. Due to the increased net charge of the phosphorylated dye (q=-4 at pH 8), it demonstrated a far better electrophoretic mobility compared with analogues with two sulfonic acid groups (q=-2). As an example, one fluorescent dye was designed to be especially convenient for practical use. It is characterized by sufficiently high chemical stability of the NHS ester, its simple isolation, handling, and solubility in aqueous buffers, as well as in organic solvents. All these features, accompanied by a zero net charge in conjugates, were accomplished by the introduction of hydrophilic groups of two types: two hydroxyl groups and one sulfonic acid residue.
Sigma-Aldrich
Phalloidin–Atto 390, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 532, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 532 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Diallyl N,N-diisopropylphosphoramidite
Sigma-Aldrich
Atto 647N azide, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N amine, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Sigma-Aldrich
Atto 425-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425-Biotin, BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Supelco
Atto 647N maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N alkyne, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N DOPE, suitable for fluorescence
Sigma-Aldrich
Atto 390 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 425, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 425 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 590-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N Protein Labeling Kit, BioReagent, suitable for fluorescence
Supelco
Atto 590 NHS ester, BioReagent, suitable for fluorescence, ≥90.0% (degree of coupling)
Sigma-Aldrich
Atto 590, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 590-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Supelco
Atto 647N, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 647N NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 390 NHS ester, BioReagent, suitable for fluorescence
Supelco
NTA - Atto 647 N, BioReagent, suitable for fluorescence, ≥80% (HPCE)
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
SAFC
Atto 465 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)