Merck
  • Evaluation of co-solvent fraction, pressure and temperature effects in analytical and preparative supercritical fluid chromatography.
Evaluation of co-solvent fraction, pressure and temperature effects in analytical and preparative supercritical fluid chromatography.
Journal of chromatography. A (2014-12-17)
Dennis Åsberg, Martin Enmark, Jörgen Samuelsson, Torgny Fornstedt
ABSTRACT
A chemometric approach is used for studying the combined effect of temperature, pressure and co-solvent fraction in analytical and preparative supercritical fluid chromatography (SFC). More specifically, by utilizing design of experiments coupled with careful measurements of the experimental conditions the interaction between pressure, temperature and co-solvent fraction was studied with respect to productivity, selectivity and retention in chiral SFC. A tris-(3,5-dimethylphenyl) carbamoyl cellulose stationary phase with carbon dioxide/methanol as mobile phase and the two racemic analytes trans-stilbene oxide (TSO) and 1,1′-bi-2-naphthol (BINOL) were investigated. It was found for the investigated model system that the co-solvent fraction and pressure were the parameters that most affected the retention factors and that the co-solvent fraction and column temperature were most important for controlling the selectivity. The productivity in the preparative mode of SFC was most influenced by the co-solvent fraction and temperature. Both high co-solvent fraction and temperature gave maximum productivity in the studied design space.
Sigma-Aldrich
1,3,5-Tri-tert-butylbenzene, 97%
Sigma-Aldrich
1,1′-Bi-2-naphthol, 99%
Sigma-Aldrich
(R)-(+)-1,1′-Bi(2-naphthol), 99%
Sigma-Aldrich
(S)-(−)-1,1′-Bi(2-naphthol), 99%
Sigma-Aldrich
Nitrous oxide, 99%