Merck
  • Molecular profiling and clinical outcome of high-grade serous ovarian cancer presenting with low- versus high-volume ascites.
Molecular profiling and clinical outcome of high-grade serous ovarian cancer presenting with low- versus high-volume ascites.
BioMed research international (2014-07-02)
Tomer Feigenberg, Blaise Clarke, Carl Virtanen, Anna Plotkin, Michelle Letarte, Barry Rosen, Marcus Q Bernardini, Alexandra Kollara, Theodore J Brown, K Joan Murphy
ABSTRACT
Epithelial ovarian cancer consists of multiple histotypes differing in etiology and clinical course. The most prevalent histotype is high-grade serous ovarian cancer (HGSOC), which often presents at an advanced stage frequently accompanied with high-volume ascites. While some studies suggest that ascites is associated with poor clinical outcome, most reports have not differentiated between histological subtypes or tumor grade. We compared genome-wide gene expression profiles from a discovery cohort of ten patients diagnosed with stages III-IV HGSOC with high-volume ascites and nine patients with low-volume ascites. An upregulation of immune response genes was detected in tumors from patients presenting with low-volume ascites relative to those with high-volume ascites. Immunohistochemical studies performed on tissue microarrays confirmed higher expression of proteins encoded by immune response genes and increased tumorinfiltrating cells in tumors associated with low-volume ascites. Comparison of 149 advanced-stage HGSOC cases with differential ascites volume at time of primary surgery indicated low-volume ascites correlated with better surgical outcome and longer overall survival. These findings suggest that advanced stage HGSOC presenting with low-volume ascites reflects a unique subgroup of HGSOC, which is associated with upregulation of immune related genes, more abundant tumor infiltrating cells and better clinical outcomes.