Merck
  • Isolation and characterization of rat intestinal bacteria involved in biotransformation of (-)-epigallocatechin.
Isolation and characterization of rat intestinal bacteria involved in biotransformation of (-)-epigallocatechin.
Archives of microbiology (2014-06-21)
Akiko Takagaki, Yuko Kato, Fumio Nanjo
ABSTRACT
Two intestinal bacterial strains MT4s-5 and MT42 involved in the degradation of (-)-epigallocatechin (EGC) were isolated from rat feces. Strain MT4s-5 was tentatively identified as Adlercreutzia equolifaciens. This strain converted EGC into not only 1-(3, 4, 5-trihydroxyphenyl)-3-(2, 4, 6-trihydroxyphenyl)propan-2-ol (1), but also 1-(3, 5-dihydroxyphenyl)-3-(2, 4, 6-trihydroxyphenyl)propan-2-ol (2), and 4'-dehydroxylated EGC (7). Type strain (JCM 9979) of Eggerthella lenta was also found to convert EGC into 1. Strain MT42 was identified as Flavonifractor plautii and converted 1 into 4-hydroxy-5-(3, 4, 5-trihydroxyphenyl)valeric acid (3) and 5-(3, 4, 5-trihydroxyphenyl)-γ-valerolactone (4) simultaneously. Strain MT42 also converted 2 into 4-hydroxy-5-(3, 5-dihydroxyphenyl)valeric acid (5), and 5-(3, 5-dihydroxyphenyl)-γ-valerolactone (6). Furthermore, F. plautii strains ATCC 29863 and ATCC 49531 were found to catalyze the same reactions as strain MT42. Interestingly, formation of 2 from EGC by strain MT4s-5 occurred rapidly in the presence of hydrogen supplied by syntrophic bacteria. Strain JCM 9979 also formed 2 in the presence of the hydrogen or formate. Strain MT4s-5 converted 1, 3, and 4 to 2, 5, and 6, respectively, and the conversion was stimulated by hydrogen, whereas strain JCM 9979 could catalyze the conversion only in the presence of hydrogen or formate. On the basis of the above results together with previous reports, the principal metabolic pathway of EGC and EGCg by catechin-degrading bacteria in gut tract is proposed.
Product Description
Supelco
(−)-Epigallocatechin, analytical standard
Supelco
(−)-Gallocatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Supelco
Acetic acid, analytical standard
Epicatechin, primary reference standard
Epigallocatechin, primary reference standard
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis