Merck
  • Structural dynamics and physicochemical properties of pDNA/DODAB:MO lipoplexes: effect of pH and anionic lipids in inverted non-lamellar phases versus lamellar phases.
Structural dynamics and physicochemical properties of pDNA/DODAB:MO lipoplexes: effect of pH and anionic lipids in inverted non-lamellar phases versus lamellar phases.
Biochimica et biophysica acta (2014-07-01)
J P Neves Silva, I M S C Oliveira, A C N Oliveira, M Lúcio, A C Gomes, P J G Coutinho, M E C D Real Oliveira
ABSTRACT
Dioctadecyldimethylammonium bromide (DODAB):Monoolein (MO) lipoplexes have mainly been studied within the range of high molar ratios of DODAB, with noticeable transfection efficiencies in the Human Embryonic Kidney (HEK, a.k.a. 293T) cell line. In this work, we intend to study the effect of high MO content on the structure and physicochemical properties of pDNA/DODAB:MO lipoplexes to achieve some correlation with their transfection efficiency. Static/Dynamic Light Scattering and Cryo-TEM imaging were used to characterize the size/morphology of DNA/DODAB:MO lipoplexes at different DODAB:MO contents (2:1, 1:1, 1:2) and charge ratios (CRs) (+/-). Nile Red fluorescence emission was performed to detect changes in microviscosity, hydration and polarity of DNA/DODAB:MO systems. Lipoplexes stability at physiological pH values and in the presence of anionic lipids was evaluated by Förster Resonance Energy Transfer (FRET). Physicochemical/structural data were complemented with transfection studies in HEK cells using the β-galactosidase reporter gene activity assay. This work reports the coexistence of multilamellar and non-lamellar inverted phases in MO-richer lipoplexes (DODAB:MO 1:2 and 1:4), leading to transfection efficiencies comparable to those of multilamellar (DODAB-richer) lipoplexes, but at higher charge ratios [CR (+/-)=6.0] and without dose-effect response. These results may be related to the structural changes of lipoplexes promoted by high MO content.
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Dimethyldioctadecylammonium bromide, ≥98% (TLC)
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Levodopa, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyldioctadecylammonium bromide, ≥98.0% (AT)
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Nile Red, Technical grade
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Nile Red, for microscopy
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Nile Red, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Glycerol mono-oleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleic acid, Selectophore, ≥99%