Merck
  • Gene-specific methylation control of H3K9 and H3K36 on neurotrophic BDNF versus astroglial GFAP genes by KDM4A/C regulates neural stem cell differentiation.
Gene-specific methylation control of H3K9 and H3K36 on neurotrophic BDNF versus astroglial GFAP genes by KDM4A/C regulates neural stem cell differentiation.
Journal of molecular biology (2014-04-22)
Anna Cascante, Susanne Klum, Moumita Biswas, Beatriz Antolin-Fontes, Fanie Barnabé-Heider, Ola Hermanson
ABSTRACT
Neural stem cell (NSC) state and fate depend on spatially and temporally synchronized transcriptional and epigenetic regulation of the expression of extrinsic signaling factors and intrinsic cell-specific genes, but the functional roles for chromatin-modifying enzymes in neural differentiation remain poorly understood. Here we show that the histone demethylases KDM4A (JMJD2A) and KDM4C (JMJD2C) are essential for proper differentiation of NSCs in vitro and in vivo. KDM4A/C were required for neuronal differentiation, survival and expression of the neurotrophic signaling factor BDNF in association with promoter H3K9 demethylation and RNA polymerase II recruitment. Unexpectedly, KDM4A/C were essential for selective H3K36 demethylation and loss of RNA polymerase II recruitment in transcribed regions of the astrocyte-characteristic gene GFAP, thereby in parallel repressing astrocytic differentiation by control of elongation. We propose that gene- and lysine-specific KDM4A/C-mediated control of histone methylation and thereby regulation of intrinsic factors and signaling factors such as BDNF provide a novel control mechanism of lineage decision.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Ciliary Neurotrophic Factor human, CNTF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Lysine acetate, European Pharmacopoeia (EP) Reference Standard