Merck
  • Regioselective copper-catalyzed amination of bromobenzoic acids using aliphatic and aromatic amines.
Regioselective copper-catalyzed amination of bromobenzoic acids using aliphatic and aromatic amines.
The Journal of organic chemistry (2006-04-08)
Christian Wolf, Shuanglong Liu, Xuefeng Mei, Adam T August, Michael D Casimir
ABSTRACT
A chemo- and regioselective copper-catalyzed cross-coupling procedure for amination of 2-bromobenzoic acids is described. The method eliminates the need for acid protection and produces N-aryl and N-alkyl anthranilic acid derivatives in up to 99% yield. N-(1-Pyrene)anthranilic acid has been employed in metal ion-selective fluorosensing. Titration experiments showed that this pyrene-derived amino acid forms an equimolar complex with Hg(II) in water resulting in selective fluorescence quenching even in the presence of other metal ions such as Zn(II) and Cd(II).