Merck
Firefly luciferase as a tool in molecular and cell biology.
Analytical biochemistry (1988-11-15)
S J Gould, S Subramani
ABSTRACT
The unique properties of firefly luciferase and the cloning of the gene for this enzyme have spawned a number of novel applications of this protein. We summarize a few of these applications including its use as a reporter gene, as a model for the study of protein import into peroxisomes, and as a component of a heterologous gene expression system.