Merck
  • Sulfonyl-containing aldophosphamide analogues as novel anticancer prodrugs targeted against cyclophosphamide-resistant tumor cell lines.
Sulfonyl-containing aldophosphamide analogues as novel anticancer prodrugs targeted against cyclophosphamide-resistant tumor cell lines.
Journal of medicinal chemistry (2004-07-09)
Monish Jain, Junying Fan, Nesrine Z Baturay, Chul-Hoon Kwon
ABSTRACT
A series of sulfonyl-group containing analogues of aldophosphamide (Aldo) were synthesized as potential anticancer prodrugs that liberate the cytotoxic phosphoramide mustards (PM, IPM, and tetrakis-PM) via beta-elimination, a nonenzymatic activation mechanism. Kinetic studies demonstrated that all these compounds spontaneously liberate phosphoramide mustards with half-lives in the range of 0.08-15.2 h under model physiological conditions in 0.08 M phosphate buffer at pH 7.4 and 37 degrees C. Analogous to Aldo, the rates of beta-elimination in all compounds was enhanced in reconstituted human plasma under same conditions. The compounds were more potent than the corresponding phosphoramide mustards against V-79 Chinese hamster lung fibroblasts in vitro (IC(50) = 1.8-69.1 microM). Several compounds showed excellent in vivo antitumor activity in CD2F1 mice against both P388/0 (Wild) and P388/CPA (CP-resistant) tumor cell lines.