Merck
  • Budding from Golgi membranes requires the coatomer complex of non-clathrin coat proteins.
Budding from Golgi membranes requires the coatomer complex of non-clathrin coat proteins.
Nature (1993-04-15)
L Orci, D J Palmer, M Ravazzola, A Perrelet, M Amherdt, J E Rothman
ABSTRACT
Do the coats on vesicles budded from the Golgi apparatus actually cause the budding, or do they simply coat buds (Fig. 1)? One view (the membrane-mediated budding hypothesis) is that budding is an intrinsic property of Golgi membranes not requiring extrinsic coat proteins. Assembly of coats from dispersed subunits is super-imposed upon the intrinsic budding process and is proposed to convert the tips of tubules into vesicles. The alternative view (the coat-mediated budding hypothesis) is that coat formation provides the essential driving force for budding. The membrane-mediated budding hypothesis was inspired by the microtubule-dependent extension of apparently uncoated, 90-nm-diameter membrane tubules from the Golgi apparatus and other organelles in vivo after treatment with brefeldin A, a drug that inhibits the assembly of coat proteins onto Golgi membranes. This hypothesis predicts that tubules will be extended when coat proteins are unavailable to convert tubule-derived membrane into vesicles. Here we use a cell-free system in which coated vesicles are formed from Golgi cisternae to show that, on the contrary, when budding diminishes as a result of immunodepletion of coat protein pools, tubules are not formed at the expense of vesicles. We conclude that coat proteins are required for budding from Golgi membranes.