Merck
  • Formation of 2-chlorobenzylidenemalononitrile (CS riot control agent) thermal degradation products at elevated temperatures.
Formation of 2-chlorobenzylidenemalononitrile (CS riot control agent) thermal degradation products at elevated temperatures.
Journal of chromatography. A (2002-06-18)
Timothy A Kluchinsky, Michael V Sheely, Paul B Savage, Philip A Smith
ABSTRACT
2-Chlorobenzylidenemalononitrile (CS riot control agent) has been shown to produce a number of thermal degradation products when dispersed at high temperature. We hypothesized that these CS-derived compounds are formed by energy input from heating during the dispersion process. Here we identified organic CS-derived compounds formed from purified CS subjected to temperatures ranging from 300 to 900 degrees C in an inert atmosphere with analysis of tube furnace effluent by gas chromatography and mass spectrometry. We conclude that the production of many CS-derived compounds previously observed during high-temperature dispersion is likely to be heat related.
Sigma-Aldrich
3-Isoquinolinecarbonitrile, 99%