Merck
  • Chemosensitivity of resistant colon cancer cell lines to lobaplatin, heptaplatin, and dicycloplatin.
Chemosensitivity of resistant colon cancer cell lines to lobaplatin, heptaplatin, and dicycloplatin.
International journal of clinical pharmacology and therapeutics (2014-07-06)
Wen Wei, Zhen Liu, Xi Chen, Feng Bi
ABSTRACT
Lobaplatin, heptaplatin, and dicycloplatin are three new platinum drugs. The aim of this study is to investigate the chemosensitivity of resistant colon cancer cell lines to these three drugs. Eight resistant colon cancer cell lines (four oxaliplatin-resistant colon cancer cell lines and four irinotecan-resistant colon cancer cell lines) were cultured in 96-well plates. Lobaplatin, heptaplatin, and dicycloplatin were added in various concentrations. IC50 was determined by MTT assay and the results were confirmed by CCK-8 assay. A cell death detection ELISA assay was performed to quantitate the apoptotic index by detecting the histone-associated DNA fragments generated by the apoptotic cells. Lobaplatin and heptaplatin had an inhibiting effect in all resistant cell lines, dicycloplatin demonstrate only weak antitumor activity in vitro. We confirmed the efficacy of two platinum drugs to oxaliplatin/ irinotecan-resistant colon cancer cell lines, suggesting these two drugs could be considered for improving the control of recurrent colon cancer following initial therapy with oxaliplatin and irinotecan.
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%