Merck
Quantification of dissolved iron sources to the North Atlantic Ocean.
Nature (2014-07-11)
Tim M Conway, Seth G John
ABSTRACT
Dissolved iron is an essential micronutrient for marine phytoplankton, and its availability controls patterns of primary productivity and carbon cycling throughout the oceans. The relative importance of different sources of iron to the oceans is not well known, however, and flux estimates from atmospheric dust, hydrothermal vents and oceanic sediments vary by orders of magnitude. Here we present a high-resolution transect of dissolved stable iron isotope ratios (δ(56)Fe) and iron concentrations ([Fe]) along a section of the North Atlantic Ocean. The different iron sources can be identified by their unique δ(56)Fe signatures, which persist throughout the water column. This allows us to calculate the relative contribution from dust, hydrothermal venting and reductive and non-reductive sedimentary release to the dissolved phase. We find that Saharan dust aerosol is the dominant source of dissolved iron along the section, contributing 71-87 per cent of dissolved iron. Additional sources of iron are non-reductive release from oxygenated sediments on the North American margin (10-19 per cent), reductive sedimentary dissolution on the African margin (1-4 per cent) and hydrothermal venting at the Mid-Atlantic Ridge (2-6 per cent). Our data also indicate that hydrothermal vents in the North Atlantic are a source of isotopically light iron, which travels thousands of kilometres from vent sites, potentially influencing surface productivity. Changes in the relative importance of the different iron sources through time may affect interactions between the carbon cycle and climate.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Iron, foil, 6mm disks, thickness 0.004mm, 99.85%
Iron, foil, 50mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.004mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.020mm, 99.85%
Iron, foil, 5m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, light tested, 50x50mm, thickness 0.038mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, 6mm disks, thickness 0.15mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.003mm, 99.85%
Iron, foil, 5m coil, thickness 0.20mm, hard, 99.5%
Iron, foil, 5m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.008mm, 99.85%
Iron, foil, 8mm disks, thickness 0.004mm, 99.85%
Iron, foil, 8mm disks, thickness 0.0125mm, 99.99+%